missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other T. Vaginalis P65 Protein

A cDNA sequence encoding the T. Vaginalis P65 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13679-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934359

  • 1665.00€ / 1mg

Estimated Shipment: 25-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

T. Vaginalis P65 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Protein is >90% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
T. Vaginalis p65protein solution in PBS and 2M Urea.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions