missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other UPP1ella Protein
A cDNA sequence encoding the UPP1ella was constructed and used to recombinantly synthesize the protein.
137.00€ - 2120.00€
Specifications
Name | UPP1ella Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Biological Activity | 30 U/mg protein. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935069
|
enQuireBio™
QP13901-10UG |
10 μg |
137.00€
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955069
|
enQuireBio™
QP13901-50UG |
50 μg |
212.00€
50µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945069
|
enQuireBio™
QP13901-1MG |
1 mg |
2120.00€
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
UPP1ella Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSKSDVFHLG LTKNDLQGAQ LAIVPGDPER VEKIAALMDK PVKLASHREF TSWRAELDGK AVIVCSTGIG GPSTSIAVEE LAQLGIRTFL RIGTTGAIQP HINVGDVLVT TASVRLDGAS LHFAPMEFPA VADFACTTAL VEAAKSIGAT THVGVTASSD TFYPGQERYD TYSGRVVRRF KGSMEEWQAM GVMNYEMESA TLLTMCASQG LRAGMVAGVI VNRTQQEIPN AETMKQTESH AVKIVVEAAR RLL | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
30 U/mg protein. | |
Other | |
Untagged | |
The UPase was lyophilized from 1 mg/ml solution containing 25mM Tris-HCl, pH 8.0, 0.15M NaCl. |