missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Uricase Protein
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Specifications
Specifications
| Name | Uricase Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | The specific activity was found to be 10U/mg. One Unit oxidizes one micromole of uric acid per minute at 25C, at pH 8.5. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | msavkaaryg kdnvrvykvh kdektgvqtv yemtvcvlle geietsytka dnsvivatds ikntiyitak qnpvtppelf gsilgthfie kynhihaahv nivchrwtrm didgkphphs firdseekrn vqvdvvegkg idiksslsgl tvlkstnsqf wgflrdeytt lketwdrils tdvdatwqwk nfsglqevrs hvpkfdatwa tarevtlktf aednsasvqa tmykmaeqil arqqlietve yslpnkhyfe idlswhkglq ntgknaevfa pqsdpnglik ctvgrsslks kl |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction