missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Y. Enterocolitica (O:8) YopM Protein
A cDNA sequence encoding the Y. Enterocolitica (O:8) YopM was constructed and used to recombinantly synthesize the protein.
910.00€ - 6055.00€
Specifications
Name | Y. Enterocolitica (O:8) YopM Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945289
|
enQuireBio™
QP13974-50UG |
50 μg |
910.00€
50µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925289
|
enQuireBio™
QP13974-100UG |
100 μg |
1365.00€
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935289
|
enQuireBio™
QP13974-1MG |
1 mg |
6055.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Y. Enterocolitica (O:8) YopM Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Y. Enterocolitica(O:8) YopM is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |