missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Y. Enterocolitica (O:9) YopH Protein

A cDNA sequence encoding the Y. Enterocolitica (O:9) YopH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13977-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15925299

  • 5535.00€ / 1mg

Estimated Shipment: 01-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Y. Enterocolitica (O:9) YopH Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
Y. Enterocolitica (O:9) YopH is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions