missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine GHBP Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | GHBP Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Ovine |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MDLWQLLLTL AVAGSSDAFS GSEATPAFFV RASQSLQILY PGLETNSSGN LKFTKCRSPE LETFSCHWTD GANHSLQSPG SVQMFYIRRD IQEWKECPDY VSAGENSCYF NSSYTSVWTP YCIKLTSNGG IVDHKCFSVE DIVQPDPPVG LNWTLLNISL TEIHADILVK WEPPPNTDVK MGWIILEYEL HYKELNETQW KMMDPLLVTS VPMYSLRLDK EYEVRVRTRQ RNTEKYGKFS EVLLITFPQM NPSACEEDFQ FPWFLIIIFG ILGLTVTLFL LIFSKQQRIK MLILPPVPVP KIKGIDPDLL KEGKLEEVNT ILAIHDNYKH EFYNDDSWVE FIELDIDDPD EKTEGSDTDR LLSNDHEKSL SIFGAKDDDS GRTSCYEPDI LETDFHVSDM CDGTSEVAQP QRLKGEADIL CLDQKNQNNS PSNDAAPASQ QPSVILVEEN KPRPLPIGGT ESTHQAVHTQ LSNPSSLANI DFYAQVSDIT PAGNVVLSPG QKNKTGNPQC DTHPEVVTPS QADFIVDSAY FCEVDAKKYI ALAPDVEAES HIEPSFNQED IYITTESLTT TAGRSGTAEN VPSSEIPVPD YTSIHIVQSP QGLVLNATAL PLPDKEFLSS CGYVSTDQLN KIMP |
| Buffer | GHBP was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction