missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ovine Leptin Protein

Product Code. 15945058
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
This item is not returnable. View return policy

Product Code. 15945058

Brand: enQuireBio™ QP1075020ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.

Specifications

Name Leptin Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
Product Type Recombinant Protein
Cross Reactivity Ovine
Species E. coli
Protein Tag Untagged
Sequence VPIRKVQDDT KTLIKTIVTR INDISHTQSV SSKQRVTGLD FIPGLHPLLS LSKMDQTLAI YQQILASLPS RNVIQISNDL ENLRDLLHLL AASKSCPLPQ VRALESLESL GVVLEASLYS TEVVALSRLQ GSLQDMLRQL DLSPGC
Buffer The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Purity or Quality Grade Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.