Learn More
enQuireBio™ Recombinant Ovine Prolactin Antagonist Protein
A cDNA sequence encoding the Prolactin Antagonist was constructed and used to recombinantly synthesize the protein.
137.00€ - 2055.00€
Specifications
Name Prolactin Antagonist Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Ovine Prolactin Antagonist is devoid of agonistic activity and capable of inhibiting biological activity of Ovine Prolactin or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells.
Product Type Recombinant Protein
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15915409
View Documents Promotion Details
enQuireBio™
QP14010-EC-10UG
10 μg
137.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
15935409
View Documents Promotion Details
enQuireBio™
QP14010-EC-50UG
50 μg
212.00€
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
15925409
View Documents Promotion Details
enQuireBio™
QP14010-EC-1MG
1 mg
2055.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Specifications
Prolactin Antagonist Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Ovine Prolactin was lyophilized from a concentrated (1 mg/ml) solution with 0.02%-0.03% NaHCO3.
Research Use Only
Ovine Prolactin Antagonist is devoid of agonistic activity and capable of inhibiting biological activity of Ovine Prolactin or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells.
Ovine
His
Greater than 99.0% as determined by SDS-PAGE.