missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ovine Prolactin Protein

Product Code. p-7148661
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15975358

Brand: enQuireBio™ QP10851EC10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Prolactin was constructed and used to recombinantly synthesize the protein.

Specifications

Name Prolactin Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Is fully biologically active as evidenced by inducing proliferation of Nb2 cells.
Product Type Recombinant Protein
Cross Reactivity Ovine
Species E. coli
Protein Tag Untagged
Sequence MDSKGSAQKG SRLLLLLVVS NLLLCQGVVS TPVCPNGPGN CQVSLRDLFD RAVMVSHYIH NLSSEMFNEF DKRYAQGKGF ITMALNSCHT SSLPTPEDKE QAQQTHHEVL MSLILGLLRS WNDPLYHLVT EVRGMKGVPD AILSRAIEIE EENKRLLEGM EMIFGQVIPG AKETEPYPVW SGLPSLQTKD EDARHSAFYN LLHCLRRDSS KIDTYLKLLN CRIIYNNNC
Buffer The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.