missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Enterokinase Protein
A cDNA sequence encoding the Enterokinase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11791-500IU
Additional Details : Weight : 0.01000kg
Specifications
397152 | |
500 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Porcine | |
MGSKRIIPSR HRSLSTYEVM FTALFAILMV LCAGLIAVSW LTIKGSEKDA ALGKSHEARG TMKITSGVTY NPNLQDKLSV DFKVLAFDIQ QMIGEIFQSS NLKNEYKNSR VLQFENGSVI VIFDLLFAQW VSDENIKEEL IQGIEANKSS QLVAFHIDVN SIDITESLEN YSTTSPSTTS DKLTTSSPPA TPGNVSIECL PGSRPCADAL KCIAVDLFCD GELNCPDGSD EDSKICATAC DGKFLLTESS GSFDAAQYPK LSEASVVCQW IIRVNQGLSI ELNFSYFNTY SMDVLNIYEG VGSSKILRAS LWLMNPGTIR IFSNQVTVTF LIESDENDYI GFNATYTAFN STELNNDEKI NCNFEDGFCF WIQDLNDDNE WERIQGTTFP PFTGPNFDHT FGNASGFYIS TPTGPGGRQE RVGLLSLPLE PTLEPVCLSF WYYMYGENVY KLSINISNDQ NIEKIIFQKE GNYGENWNYG QVTLNETVEF KVAFNAFKNQ FLSDIALDDI SLTYGICNVS LYPEPTLVPT SPPELPTDCG GPFELWEPNT TFTSMNFPNN YPNQAFCVWN LNAQKGKNIQ LHFEEFDLEN IADVVEIRDG EEDDSLLLAV YTGPGPVEDV FSTTNRMTVL FITNDALTKG GFKANFTTGY HLGIPEPCKE DNFQCENGEC VLLVNLCDGF SHCKDGSDEA HCVRFLNGTA NNSGLVQFRI QSIWHTACAE NWTTQTSDDV CQLLGLGTGN SSMPFFSSGG GPFVKLNTAP NGSLILTASE QCFEDSLILL QCNHKSCGKK QVAQEVSPKI VGGNDSREGA WPWVVALYYN GQLLCGASLV SRDWLVSAAH CVYGRNLEPS KWKAILGLHM TSNLTSPQIV TRLIDEIVIN PHYNRRRKDS DIAMMHLEFK VNYTDYIQPI |
Enterokinase Protein | |
Research Use Only | |
TMPRSS15 | |
Porcine | |
Untagged | |
2 IU/microL, 50mM Tris-HCl, pH 8.0, 0.5M NaCl and 50% glycerol. |