missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine OXM Protein
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
Specifications
Specifications
| Name | OXM Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Porcine |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-Lys-Arg-Asn-Lys-Asn-Asn-Ile-Ala |
| Buffer | Lyophilized from a concentrated (1 mg/ml) solution in water containing no additives. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction