missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Thrombin Protein
Click to view available options
Quantity:
1000 IU
2000 IU
500 U
Unit Size:
1000 units
2000 units
500 units
Specifications
Specifications
| Name | Thrombin Protein |
| Quantity | 500 U |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | One unit is defined as the amount of enzyme needed to cleave 1 mg of fusion protein in 16 hours to 95% completion at 20C in a buffer containing 25mM Tris-HCl, pH 8.4, 150mM NaCl, and 2.5mM CaCl2. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Porcine |
| Species | Porcine |
| Protein Tag | Untagged |
| Buffer | Lyophilized Powder from 20mM PBS, pH 7.4 containing 0.9% NaCl. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction