missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CALB1 Protein

A cDNA sequence encoding the CALB1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11257-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15926638

  • 125.00€ / 1µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

83839
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Calb1
Rat
Untagged
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 50mM NaHCO3.
CALB1 Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MAESHLQSSL ITASQFFEIW LHFDADGSGY LEGKELQNLI QELLQARKKA GLELSPEMKT FVDQYGQRDD GKIGIVELAH VLPTEENFLL LFRCQQLKSC EEFMKTWRKY DTDHSGFIET EELKNFLKDL LEKANKTVDD TKLAEYTDLM LKLFDSNNDG KLELTEMARL LPVQENFLLK FQGIKMCGKE FNKAFELYDQ DGNGYIDENE LDALLKDLCE KNKQELDINN ISTYKKNIMA LSDGGKLYRT DLALILSAGD N
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions