missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat Cardiotrophin-1/CT-1 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
Cardiotrophin 1 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.5 ng/mL, corresponding to a specific activity of > 2.0 x 10^6 IU/mg.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 30 % Acetonitrile, 0.1 % TFA. |
| Gene ID (Entrez) | 1489 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 21.4 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of Cardiotrophin-1/CT-1 AS determined by LAL method. |
| Gene Alias | cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1 CT1 |
| Gene Symbol | CTF1 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in 4 mM HCl to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction