missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CD74 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) CD74, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5796-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037201.1 | |
CD74 Protein | |
Research Use Only | |
Cd74 | |
Recombinant Protein | |
E. coli | |
QQQGRLDKLT VTSQNLQLEN LRMKLPKSAK PVSPMRMATP LLMRPLSMDN MLQAPVKNVT KYGNMTQDHV MHLLTKSGPV NYPQLKGSFP ENLKHLKNSM NGLDWKVFES WMKQWLLFEM SKNSLEEKQP TQTPPKVLTK CQEEVSHIPD VHPGAFRPKC DENGNYMPLQ CHGSTGYCWC VFPNGTEVPH TKSRGRHNCS EPLDMEDPSS GLGVTKQDMG QMFL | |
Greater than 90% as determined by SDS-PAGE. |
25599 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |