missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat Defensin beta 1 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
Defensin beta 1 protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using CD34+ dendritic cells is in a concentration range of 100.0-1000.0 ng/mL
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, 500 mM NaCl, pH 7.0. |
| Gene ID (Entrez) | 1672 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 4.1 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of Defensin beta 1 as determined by LAL method. |
| Gene Alias | BD1, BD-1, beta-defensin 1, beta-defensin-1, DEFB-1, DEFB101, defensin, beta 1 HBD-1, hBD-1, HBD1MGC51822 |
| Gene Symbol | DEFB1 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction