missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FGF 21 Protein
A cDNA sequence encoding the FGF 21 was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Name | FGF 21 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50 determined by a cell proliferation assay using murine NIH/3T3 cells is less than 700 ng/ml, corresponding to a specific activity of > 1.4 M 1000 IU/mg in the presence of 5microg/ml of rMuKlotho-beta. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934378
|
enQuireBio™
QP10600-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924378
|
enQuireBio™
QP10600-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914378
|
enQuireBio™
QP10600-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FGF 21 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
AYPISDSSPL LQFGGQVRQR YLYTDDDQDT EAHLEIREDG TVVGTAHRSP ESLLELKALK PGVIQILGVK ASRFLCQQPD GTLYGSPHFD PEACSFRELL LKDGYNVYQS EAHGLPLRLP QKDSQDPATR GPVRFLPMPG LPHEPQEQPG VLPPEPPDVG SSDPLSMVEP LQGRSPSYAS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The ED50 determined by a cell proliferation assay using murine NIH/3T3 cells is less than 700 ng/ml, corresponding to a specific activity of > 1.4 M 1000 IU/mg in the presence of 5microg/ml of rMuKlotho-beta. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4. |