missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Guanidinoacetate N-methyltransferase Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Guanidinoacetate N-methyltransferase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6068-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959749

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Specifications

Specifications

Guanidinoacetate N-methyltransferase Protein
Research Use Only
Gamt
Recombinant Protein
E. coli
MSSSAASPLF APGEDCGPAW RAAPAAYDTS DTHLQILGKP VMERWETPYM HSLAAAAASR GGRVLEVGFG MAIAASRVQQ APIKEHWIIE CNDGVFQRLQ NWALKQPHKV VPLKGLWEEE APTLPDGHFD GILYDTYPLS EETWHTHQFN FIKTHAFRLL KPGGILTYCN LTSWGELMKS KYTDITAMFE ETQVPALLEA GFQRENICTE VMALVPPADC RYYAFPQMIT PLVTKH
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions