missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Guanidinoacetate N-methyltransferase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Guanidinoacetate N-methyltransferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6068-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
Guanidinoacetate N-methyltransferase Protein | |
Research Use Only | |
Gamt | |
Recombinant Protein | |
E. coli | |
MSSSAASPLF APGEDCGPAW RAAPAAYDTS DTHLQILGKP VMERWETPYM HSLAAAAASR GGRVLEVGFG MAIAASRVQQ APIKEHWIIE CNDGVFQRLQ NWALKQPHKV VPLKGLWEEE APTLPDGHFD GILYDTYPLS EETWHTHQFN FIKTHAFRLL KPGGILTYCN LTSWGELMKS KYTDITAMFE ETQVPALLEA GFQRENICTE VMALVPPADC RYYAFPQMIT PLVTKH | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |