missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IFN-gamma Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and PEG with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in Tris, NaCl and PEG 3350.
Unit Size:
100µg
Description
The Recombinant Rat IFN-gamma Protein is derived from E. coli. The Recombinant Rat IFN-gamma Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl and PEG 3350. |
| Gene ID (Entrez) | 25712 |
| Molecular Weight (g/mol) | 15.5 kDa |
| Name | IFN-gamma |
| Quantity | 100 μg |
| Source | E. coli-derived rat IFN-gamma protein Glu23-Cys156,with an N-terminal Met |
| Storage Requirements | 3 months from date of receipt, 2 to 8° C as supplied. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction