missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IFNG / Interferon Gamma Protein

Recombinant Protein

112.00€ - 1665.00€

Specifications

Accession Number NP_620235.1
Gene ID (Entrez) 25712
Name IFNG / Interferon Gamma Protein
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15906259
View Documents Promotion Details
enQuireBio™
QP5496-20UG
20 μg
112.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15986249
View Documents Promotion Details
enQuireBio™
QP5496-100UG
100 μg
263.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15916259
View Documents Promotion Details
enQuireBio™
QP5496-250UG
250 μg
580.00€
250µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15926259
View Documents Promotion Details
enQuireBio™
QP5496-500UG
500 μg
985.00€
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15996249
View Documents Promotion Details
enQuireBio™
QP5496-1MG
1 mg
1665.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_620235.1
IFNG / Interferon Gamma Protein
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MQGTLIESLE SLKNYFNSSS MDAMEGKSLL LDIWRNWQKD GNTKILESQI ISFYLRLFEV LKDNQAISNN ISVIESHLIT NFFSNSKAKK DAFMSIAKFE VNNPQIQHKA VNELIRVIHQ LSPESSLRKR KRSRC
Greater than 95% as determined by SDS-PAGE.
25712
Research Use Only
IFNG
Rat
Untagged
10 mM sodium phosphate, 100 mM sodium chloride, pH 7.5
Videos
SDS
Documents

Documents

Product Certifications