missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 13, Partial Protein

A cDNA sequence encoding the IL 13, Partial was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP14019-EC-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15925429

  • 6480.00€ / 1mg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

116553
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Il13
Recombinant Protein
E. coli
MALWVTAVLA LACLGGLATP GPVRRSTSPP VALRELIEEL SNITQDQKTS LCNSSMVWSV DLTAGGFCAA LESLTNISSC NAIHRTQRIL NGLCNQKASD VASSPPDTKI EVAQFISKLL NYSKQLFRYG H
IL 13, Partial Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
ED50 range = 2-6 ng/mL as determined by the dose dependent proliferation of human TF-1 cells. Optimal concentration for individual application should be determined by a dose response assay.
Rat
Untagged
The protein (1 mg/ml) was lyophilized with no additives.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions