missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 13, Partial Protein

Product Code. 15935429
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15935429

Brand: enQuireBio™ QP14019EC2ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IL 13, Partial was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 116553
Name IL 13, Partial Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol Il13
Biological Activity ED50 range = 2-6 ng/mL as determined by the dose dependent proliferation of human TF-1 cells. Optimal concentration for individual application should be determined by a dose response assay.
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag Untagged
Sequence MALWVTAVLA LACLGGLATP GPVRRSTSPP VALRELIEEL SNITQDQKTS LCNSSMVWSV DLTAGGFCAA LESLTNISSC NAIHRTQRIL NGLCNQKASD VASSPPDTKI EVAQFISKLL NYSKQLFRYG H
Buffer The protein (1 mg/ml) was lyophilized with no additives.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.