Learn More
enQuireBio™ Recombinant Rat IL 13, Partial Protein
A cDNA sequence encoding the IL 13, Partial was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP14019-EC-10ug
Additional Details : Weight : 0.01000kg
Specifications
116553 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
ED50 range = 2-6 ng/mL as determined by the dose dependent proliferation of human TF-1 cells. Optimal concentration for individual application should be determined by a dose response assay. | |
Rat | |
Untagged | |
The protein (1 mg/ml) was lyophilized with no additives. |
IL 13, Partial Protein | |
Research Use Only | |
Il13 | |
Recombinant Protein | |
E. coli | |
MALWVTAVLA LACLGGLATP GPVRRSTSPP VALRELIEEL SNITQDQKTS LCNSSMVWSV DLTAGGFCAA LESLTNISSC NAIHRTQRIL NGLCNQKASD VASSPPDTKI EVAQFISKLL NYSKQLFRYG H | |
Greater than 95% as determined by SDS-PAGE. |