missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat ITGB1 / Integrin beta-1 / CD29 Protein

Product Code. 16091860
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16091860

Brand: enQuireBio™ QP6242ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) ITGB1 / Integrin beta-1 / CD29, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_058718.2
Gene ID (Entrez) 24511
Name ITGB1 / Integrin beta-1 / CD29 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Itgb1
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag His-SUMO
Sequence QTDKNRCLKA NAKSCGECIQ AGPNCGWCTN TTFLQEGMPT SARCDDLEAL KKKGCHPSDI ENPRGSQTIK KNKNVTNRSK GMAEKLRPED ITQIQPQQLL LKLRSGEPQK FTLKFKRAED YPIDLYYLMD LSYSMKDDLE NVKSLGTDLM NEMRRITSDF RIGFGSFVEK TVMPYISTTP AKLRNPCTSE QNCTSPFSYK NVLSLTDRGE FFNELVGQQR ISGNLDSPEG GFDAIMQVAV CGSLIGWRNV TRLLVFSTDA GFHFAGDGKL GGIVLPNDGQ CHLENNVYTM SHYYDYPSIA HLVQKLSENN IQTIFAVTEE FQPVYKELKN LIPKSAVGTL SGNSSNVIQL IIDAYNSLSS EVILENSKLP DGVTINYKSY CKNGVNGTGE NGRKCSNISI GDEVQFEISI TANKCPNKES ENQLKLNPLG FTEEVEVVLQ FICKCNCQSH GIPASPKCHE GNGTFECGAC RCNEGRVGRH CECSTDEVNS EDMDAYCRKE NSSEICSNNG ECVCGQCVCR KRENTNEIYS GKFCECDNFN CDRSNGLICG GNGVCRCRVC ECYPNYTGSA CDCSLDTVPC VATNGQICNG RGICECGACK CTDPKFQGPT CETCQTCLGV CAEHKECVQC RAFNKGEKKD TCAQECSHFN LTKVESREKL PQPVQVDPVT HCKEKDIDDC WFYFTYSVNS KGEAHVHVVE TPDCPTGPD
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.