missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat NME2 Protein

A cDNA sequence encoding the NME2 was constructed and used to recombinantly synthesize the protein.

137.00€ - 4090.00€

Specifications

Name NME2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Rat
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15921759
View Documents Promotion Details
enQuireBio™
QP12854-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15911759
View Documents Promotion Details
enQuireBio™
QP12854-20UG
20 μg
212.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15901759
View Documents Promotion Details
enQuireBio™
QP12854-1MG
1 mg
4090.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NME2 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
His
NME2 protein solution (1 mg/ml) containing Phosphate Buffered Saline pH 7.4 and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMANLERT FIAIKPDGVQ RGLVGEIIKR FEQKGFRLVA MKFLRASEEH LKQHYIDLKD RPFFPGLVKY MNSGPVVAMV WEGLNVVKTG RVMLGETNPA DSKPGTIRGD FCIQVGRNII HGSDSVESAE KEIGLWFKPE ELIDYKSCAH DWVYE
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications