missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat PD-L1/B7-H1 Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. Immobilized Recombinant Rat PD-1 Fc Chimera (Catalog # 9814-PD ) at 1 μg/mL (100 μL/well) can bind Recombinant Rat PD-L1/B7-H1 Fc Chimera with an ED50 of 0.09-0.54 μg/mL.
Specifications
Specifications
| Accession Number | NP_001178883 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 29126 |
| Molecular Weight (g/mol) | M.W. (Observed): 74-84 kDa, reducing conditions; M.W. (theoretical): 52 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived rat PD-L1/B7-H1 protein Rat B7-H1 (Phe19-Thr238) (N-terminus) Accession # NP_001178883 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction