missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SDF 1b Protein
A cDNA sequence encoding the SDF 1b was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Gene ID (Entrez) | 24772 |
---|---|
Name | SDF 1b Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Cxcl12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955398
|
enQuireBio™
QP10863-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935398
|
enQuireBio™
QP10863-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945398
|
enQuireBio™
QP10863-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
24772 | |
Research Use Only | |
Cxcl12 | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKSNNRQVC IDPKLKWIQE YLDKALNKRL KM | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
SDF 1b Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity as determined by its ability to chemoattract human peripheral blood monocytes at 100-150ng/ml corresponding to a Specific Activity of 6,667-10,000IU/mg. | |
Rat | |
Untagged | |
The Rat SDF1B protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing 20mM Phsophate buffer pH 7.4 and 0.15M NaCl. |