missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rubella Virus Rubella E2 Protein
A cDNA sequence encoding the Rubella E2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13374-500ug
Additional Details : Weight : 0.01000kg
Specifications
Rubella Virus Rubella E2 Protein | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
ubella Virus | |
Untagged |
Rubella protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM imidazol, 8M urea and 0.3M NaCl. |