missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salamander Leptin Salamander Protein
A cDNA sequence encoding the Leptin Salamander was constructed and used to recombinantly synthesize the protein.
402.00€ - 1815.00€
Specifications
Name | Salamander Leptin Salamander Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | By inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its activity in that test is similar to that of mouse leptin (see Salomon et al (2006) Protein Expression and Purifcation 47, 128–136). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15915088
|
enQuireBio™
QP10760-100UG |
100 μg |
402.00€
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935088
|
enQuireBio™
QP10760-200UG |
200 μg |
530.00€
200µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925088
|
enQuireBio™
QP10760-1MG |
1 mg |
1815.00€
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Salamander Leptin Salamander Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The sequence of the ten N-terminal amino acids was determined and was found to be Ala-Ile-Met-Val-Asp-Gln-Leu-Arg-Met-Asp | |
Greater than 99.0% as determined by: (a) Analysis by Gel-Filtration. (b) Analysis by SDS-PAGE. |
Research Use Only | |
By inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its activity in that test is similar to that of mouse leptin (see Salomon et al (2006) Protein Expression and Purifcation 47, 128–136). | |
Salamander | |
Untagged | |
The Salamander Leptin was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |