missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Salamander Leptin Salamander Protein

Artikelnummer. 15925088
Klik for at se tilgængelige muligheder
Quantity:
1 mg
100 μg
200 μg
Pakningsstørrelse:
100µg
1mg
200µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 15925088

Brand: enQuireBio™ QP107601mg

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

A cDNA sequence encoding the Leptin Salamander was constructed and used to recombinantly synthesize the protein.

Tekniske data

Name Salamander Leptin Salamander Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity By inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its activity in that test is similar to that of mouse leptin (see Salomon et al (2006) Protein Expression and Purifcation 47, 128–136).
Product Type Recombinant Protein
Cross Reactivity Salamander
Species E. coli
Protein Tag Untagged
Sequence The sequence of the ten N-terminal amino acids was determined and was found to be Ala-Ile-Met-Val-Asp-Gln-Leu-Arg-Met-Asp
Buffer The Salamander Leptin was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Purity or Quality Grade Greater than 99.0% as determined by: (a) Analysis by Gel-Filtration. (b) Analysis by SDS-PAGE.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.