missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salmonella Typhi S. Typhi OMP Protein
A cDNA sequence encoding the S. Typhi OMP was constructed and used to recombinantly synthesize the protein.
238.00€ - 1815.00€
Specifications
Name | Salmonella Typhi S. Typhi OMP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Salmonella Typhi |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15913399
|
enQuireBio™
QP13384-100UG |
100 μg |
238.00€
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933399
|
enQuireBio™
QP13384-500UG |
500 μg |
910.00€
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923399
|
enQuireBio™
QP13384-1MG |
1 mg |
1815.00€
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Salmonella Typhi S. Typhi OMP Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Salmonella Typhi | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing 10mM Tris-HCl, 1mM EDTA and 50mM arginine. |