missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant TEV Protease Protein
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
Description
Measured by its ability to cleave a fusion protein containing the recognition sequence Glu-Asn-Leu-Tyr-Phe-Gln, with the cleavage point after Gln. TEV Protease cleaves =50% of the control substrate, as measured under the described conditions. It is recommended that the cleavage for each fusion protein be optimized by varying the amount of Recombinant Viral TEV Protease, reaction time, or incubation temperature.
Specifications
Specifications
| Accession Number | NP_062908 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, EDTA, DTT and Glycerol. |
| Gene ID (Entrez) | 1502321 |
| Molecular Weight (g/mol) | M.W. (Observed): 25-30 kDa, reducing conditions; M.W. (theoretical): 28.5 kDa |
| Name | Sleeping Beauty Transposase |
| Quantity | 200 μg |
| Source | E. coli-derived viral TEV Protease protein Ala TEV Protease Glu2039-Gln2279 (Ser2256Asn) (N-terminus) Accession # NP_062908 Leu 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction