missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Varicella Zoster Virus VZV gE Protein
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Specifications
Specifications
| Name | Varicella Zoster Virus VZV gE Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Varicella Zoster Virus |
| Species | E. coli |
| Protein Tag | GST |
| Buffer | 60mM NaCl, 50mM Tris-HCl pH 8.0, 0.25% Sarkosil, 10mM glutathione and 50% glycerol. |
| Purity or Quality Grade | Varicella protein is >90% pure as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction