missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Varicella Zoster Virus VZV gE Protein
A cDNA sequence encoding the VZV gE was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13950-500ug
Additional Details : Weight : 0.01000kg
Specifications
Varicella Zoster Virus VZV gE Protein | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Varicella Zoster Virus | |
GST |
Varicella protein is >90% pure as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
60mM NaCl, 50mM Tris-HCl pH 8.0, 0.25% Sarkosil, 10mM glutathione and 50% glycerol. |