missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus HTNV Protein
A cDNA sequence encoding the HTNV was constructed and used to recombinantly synthesize the protein.
238.00€ - 1815.00€
Specifications
Name | Virus HTNV Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15910169
|
enQuireBio™
QP12359-100UG |
100 μg |
238.00€
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930169
|
enQuireBio™
QP12359-500UG |
500 μg |
910.00€
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920169
|
enQuireBio™
QP12359-1MG |
1 mg |
1815.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Virus HTNV Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
GST | |
HTNV protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HTNV (5 mg/ml) is formulated in 1xPBS pH 7.4 and 0.5mM EDTA. |