missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Norovirus Group-2 P-Domain Protein
A cDNA sequence encoding the Norovirus Group-2 P-Domain was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12868-500ug
Additional Details : Weight : 0.01000kg
Specifications
Virus Norovirus Group-2 P-Domain Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffer and 17mM K2CO3. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |