missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Yeast IDH1 Protein

Product Code. 15924598
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 15924598

Brand: enQuireBio™ QP106705mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IDH1 was constructed and used to recombinantly synthesize the protein.

Specifications

Name IDH1 Protein
Quantity 5 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The specific activity was found to be 115 U/mg.
Product Type Recombinant Protein
Cross Reactivity Yeast
Species Yeast
Protein Tag Untagged
Buffer One ml of solution contains 0.075 mol/l KPO4, 50% Glycerol, pH 7.1.
Purity or Quality Grade Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.