missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zika Ectodomain Protein

A cDNA sequence encoding the Zika Ectodomain was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13996-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935359

  • 2490.00€ / 100µg

Estimated Shipment: 28-06-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Zika Ectodomain Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Zika
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
The Zika Ectodomain solution 10mM Sodium phosphate, pH 7.2, 150mM NaCl.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions