Learn More
Deutsch Neumann™ Vacuum Quality Laboratory Tubing
Click to view available options
Diameter (Metric) Inner:
8 mm
10 mm
Diameter (Metric) Outer:
16 mm
20 mm
Overall Length (Metric):
25 m
Unit Size:
1m
25m
Description
- Durometer 50 Shore A
- Good elastic memory
- Excellent choice for vacuum applications
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2008 |
| Diameter (Metric) Inner | 10 mm |
| Material | Natural Rubber |
| Diameter (Metric) Outer | 20 mm |
| Overall Length (Metric) | 25 m |
| Thickness (Metric) | 5 mm |
| For Use With (Application) | For Laboratory Use |
| Color | Red |