missing translation for 'onlineSavingsMsg'
Learn More

Verenigde Kunststof Bedrijven™ Polypropylene Closure for Plastic Bottles

Product Code. 10470265
Click to view available options
Thread Size (Metric):
40 mm
80 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10470265

Brand: Verenigde Kunststof Bedrijven™ 407602

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Verenigde Kunststof Bedrijven™ Polypropylene Closure for Plastic Bottles features a white foam insert to offer a tight seal when working with plastic bottles.

  • PP Closure
  • Color RED
  • Food-grade
  • Foam insert

Specifications

Color Red
Material Polypropylene
No. per Pack 1 Each
Quantity 1 Each
Thread Size (Metric) 40 mm
Closure Color Red
Product Type Screw Cap
For Use With (Equipment) For white bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.