missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Reference™ 2 BASIC - Trade-In Pipette
GREENER_CHOICE

Product Code. 15743489 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Color:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15743489

Brand: Eppendorf™ 4924000215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Minimize risk of aerosol contamination and reduce thumb strain with volume pipettes

  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Ergonomic handling with reduced operating effort
  • Simple and quick tip ejection with active aerosol reduction
  • Reduce repetitive strain injuries by not having to move your thumb sideways
  • High precision and accuracy provides reliable pipetting results
TRUSTED_SUSTAINABILITY

Specifikationer

Certifications/Compliance Lot Specific Certificate
Compatible Tips epT.I.P.S.
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Dark Gray
Disposable Reusable
For Use With epT.I.P.S. 10 μL
Grip Type Ergonomic
Material Plastic
Number of Channels 1
Programmability No
Sterility Non-sterile
Type Manual Pipette
Volume (Metric) 0.1 to 2.5 μL
For Use With (Application) Pipette Tips
For Use With (Equipment) Pipette Tips
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.