missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 4.005, Chem-Lab

Product Code. 11933353
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11933353

Brand: Chem-Lab CL07.0204.0500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for pH measurement and maintenance. Buffer Solution, pH 4.005, Chem-Lab is a solution of 10.13g potassium hydrogen phthalate per 1L water, with a pH level of 4.005 ±0.01 at 25°C. NIST traceable solution is manufactured according to Eur. Ph. 4(2.2.3) standards.

pH Levels ± 0.01:
  • 0°C—pH 4.005
  • 5°C—pH 4.003
  • 10°C—pH 4.001
  • 15°C—pH 4.002
  • 20°C—pH 4.003
  • 25°C—pH 4.005
  • 30°C—pH 4.010
  • 35°C—pH 4.020
  • 37°C—pH 4.025
  • 40°C—pH 4.033
  • 45°C—pH 4.047
  • 50°C—pH 4.061

Specifications

Chemical Name or Material Potassium hydrogen phthalate
CAS 877-24-7, 7732-18-5
Melting Point 0°C
Boiling Point 100°C
For Use With (Application) Laboratory use
pH 4.005 ±0.01
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Formulation 10.13g C8H5KO4 per 1L H2O
Molecular Formula C8H5KO4
Odor Odorless
Traceability to NIST Yes (SRM - 185g)
Solubility Information Soluble in water
Formula Weight 201.23
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.