Learn More
Fisherbrand REFERENCE ELECTRODE POROUS POLYMERjunction gel filled, Calomel rugged epoxy body
Brand: Fisherbrand 93X218910
REFERENCE ELECTRODE POROUS POLYMERjunction gel filled, Calomel rugged epoxy body