Learn More
Abeba™ Reflexor™ 31064 Shoes
ESD dissipative shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 31064/37
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 15638477
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Smooth leather white/black
  • Slip resistance PU outsole sole
  • Integrated Reflexor™ massage footbed
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • velcro fastening
  • Hand-sewn seam
Specifications
Specifications
EN 61340
Microfiber
37
Polyurethane
White
Leather
Slip-resistant
31064