Learn More
Abeba™ Reflexor™ 31064 Shoes
ESD dissipative shoe includes PU-sole with high slip resistance.
Brand: Abeba™ 31064/38
Additional Details : Poids : 0.70000kg
Description
- Smooth leather white/black
- Slip resistance PU outsole sole
- Integrated Reflexor™ massage footbed
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Instep area with elastic band
- velcro fastening
- Hand-sewn seam
Specifications
EN 61340 | |
Microfiber | |
38 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
31064 |