Learn More
Abeba™ Reflexor™ 31094 Shoes
ESD dissipative shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 31094/36
Additional Details : Weight : 0.55000kg
 View more versions of this product
Product Code. 15618517
  • 90.95€ / 1 pair
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • Smooth leather white
  • Slip resistance PU outsole sole
  • Integrated Reflexor™ massage footbed
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Hand-sewn seam
Specifications
Specifications
EN 61340
Microfiber
36
Polyurethane
White
Leather
Slip-resistant
31094