Learn More
Abeba™ Reflexor™ 5710 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 5710/43
Additional Details : Weight : 1.20000kg
 View more versions of this product
Product Code. 15681918
  • 137.25€ / 1 pair
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 SRB
  • Smooth leather
  • Black
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3566/ 3570)
  • Antistatic
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 O1 SRB
Microfiber
43
Polyurethane
Black
Leather
Slip-resistant
5710