Learn More
Abeba™ Reflexor™ 6902 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand: Abeba™ 6902/42
Additional Details : Weight : 0.85000kg
Description
- CE EN ISO 20347:2012 OB A, E SRC
- Smooth leather white
- Slip resistance PU outsole sole
- 2 replaceable Reflexor™ insoles (article no. 3565/ 3567)
- Antistatic
- Lining microfiber, respiration absorbing
- 3 adjustable instep straps
- velcro fastening
- Hand-sewn seam
Specifications
CE EN ISO 20347:2012 OB A, E SRC | |
Microfiber | |
42 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
6902 |