Learn More
Abeba™ Reflexor™ 6903 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 6903/37
Additional Details : Weight : 0.85000kg
 View more versions of this product
Product Code. 15622418
  • 126.12€ / 1 pair
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather white
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3565/ 3567)
  • Antistatic
  • Heel strap foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRB
Microfiber
37
Polyurethane
White
Leather
Slip-resistant
6903